Tetrafluorhydrochinon, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2,3,5,6-Tetrafluorohydroquinone, Tetrafluorobenzene-1,4-diol

M-Clarity™ Qualitätsstufe = MQ100

104361-5G
SIAL104361-5GEA 69.3 EUR
SIAL104361-5G
Tetrafluorhydrochinon, Sigma-Aldrich®
Tetrafluorhydrochinon
Formel: C₆H₂F₄O₂
Molecular Weight: 182,07 g/mol
MDL: MFCD00002340
CAS-Nummer: 771-63-1
EINECS: 212-237-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR