Tetrafluorterephthalonitril, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Tetrafluor-1,4-benzoldicarbonitril, Tetrafluorbenzol-1,4-dicarbonitril, Tetrafluor-1,4-dicyanobenzol, Tetrafluorterephthalsäuredinitril, Tetrafluor-1,4-dicyanbenzol
M-Clarity™ Qualitätsstufe = MQ100
104426-5G
SIAL104426-5GEA
31.5
EUR
SIAL104426-5G
Tetrafluorterephthalonitril, Sigma-Aldrich®
Tetrafluorterephthalonitril
Formel:
C₈F₄N₂ Molecular Weight: 200,1 g/mol |
MDL:
MFCD00001776 CAS-Nummer: 1835-49-0 EINECS: 217-397-3 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...