4-Methoxybenzaldehyddimethylacetal, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
1-(Dimethoxymethyl)-4-methoxybenzol, 4-Methoxy-α,α-dimethoxytoluol, p-Anisaldehyddimethylacetal, p-(Dimethoxymethyl)anisol
10445-50ML-F
SIAL10445-50ML-FEA
121
EUR
SIAL10445-50ML-F
4-Methoxybenzaldehyddimethylacetal, Sigma-Aldrich®
4-Methoxybenzaldehyddimethylacetal
Formel:
C₁₀H₁₄O₃ Molecular Weight: 182,22 g/mol |
MDL:
MFCD00036507 CAS-Nummer: 2186-92-7 EINECS: 218-577-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...