1H-1,2,4-Triazol-3-thiol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3-Mercapto-1H-1,2,4-triazol, 3-Mercapto-1,2,4-triazol

M-Clarity™ Qualitätsstufe = MQ100

104558-10G
SIAL104558-10GEA 28 EUR
SIAL104558-10G
1H-1,2,4-Triazol-3-thiol, Sigma-Aldrich®
1H-1,2,4-Triazol-3-thiol
Formel: C₂H₃N₃S
Molecular Weight: 101,13 g/mol
Schmelzpunkt: 217…220 °C
MDL: MFCD00005229
CAS-Nummer: 3179-31-5
EINECS: 221-656-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR