(±)-1,1′-Bi-2-naphthol, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ200

104655-25G 104655-5G
SIAL104655-25GEA 116 EUR
SIAL104655-25G SIAL104655-5G
(±)-1,1′-Bi-2-naphthol, Sigma-Aldrich®
(±)-1,1'-Bi-2-naphthol
Formel: C₂₀H₁₄O₂
MDL: MFCD00004068
CAS-Nummer: 602-09-5
EINECS: 210-014-0
UN: 2811
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR