Phenyltrimethoxysilan, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

104744-250ML 104744-50ML
SIAL104744-250MLEA 130 EUR
SIAL104744-250ML SIAL104744-50ML
Phenyltrimethoxysilan, Sigma-Aldrich®
Phenyltrimethoxysilan
Formel: C₉H₁₄O₃Si
Dichte: 1,064 g/cm³ (20 °C)
MDL: MFCD00025689
CAS-Nummer: 2996-92-1
EINECS: 221-066-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR