Chlor(methyl)diphenylsilan, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

104760-25G
SIAL104760-25GEA 175 EUR
SIAL104760-25G
Chlor(methyl)diphenylsilan, Sigma-Aldrich®
Chlor(methyl)diphenylsilan
Formel: C₁₃H₁₃ClSi
Molecular Weight: 232,78 g/mol
MDL: MFCD00000498
CAS-Nummer: 144-79-6
EINECS: 205-639-0
UN: 2987
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR