2-(Bromacetyl)naphthalin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: α-Bromo-2'-acetonaphthone, 2-Bromo-2'-acetonaphthone

M-Clarity™ Qualitätsstufe = MQ100

105120-5G
SIAL105120-5GEA 46.6 EUR
SIAL105120-5G
2-(Bromacetyl)naphthalin, Sigma-Aldrich®
2-(Bromacetyl)naphthalin
Formel: C₁₂H₉BrO
Molecular Weight: 249,11 g/mol
MDL: MFCD00004109
CAS-Nummer: 613-54-7
EINECS: 210-348-7
UN: 3261
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR