Natrium-1-hexadecansulfonat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Sodium hexadecane-1-sulphonate, Hexadecane-1-sulphonic acid sodium salt

106410-10G
SIAL106410-10GEA 82.2 EUR
SIAL106410-10G
Natrium-1-hexadecansulfonat, Sigma-Aldrich®
Natrium-1-hexadecansulfonat
Formel: C₁₆H₃₃NaSO₃
Molecular Weight: 328,49 g/mol
MDL: MFCD00007528
CAS-Nummer: 15015-81-3
EINECS: 239-100-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR