meso-1,2-Dibrom-1,2-diphenylethan, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

106550-100G 106550-25G
SIAL106550-100GEA 255 EUR
SIAL106550-100G SIAL106550-25G
meso-1,2-Dibrom-1,2-diphenylethan, Sigma-Aldrich®
meso-1,2-Dibrom-1,2-diphenylethan
Formel: C₁₄H₁₂Br₂
MDL: MFCD00000137
CAS-Nummer: 13440-24-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR