3-Nitro-o-kresol, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
1-Hydroxy-2-methyl-3-nitrobenzol, 2-Methyl-3-nitrophenol, 2-Hydroxy-6-nitrotoluol
M-Clarity™ Qualitätsstufe = MQ100
106941-10G
SIAL106941-10GEA
174
EUR
SIAL106941-10G
3-Nitro-o-kresol, Sigma-Aldrich®
3-Nitro-o-kresol
Formel:
C₇H₇NO₃ Molecular Weight: 153,14 g/mol Schmelzpunkt: 149…152 °C |
MDL:
MFCD00007241 CAS-Nummer: 5460-31-1 EINECS: 226-739-0 UN: 2446 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...