1-Brom-2,3,4,5-tetrafluorbenzol, Sigma-Aldrich®
Lieferant: Merck
|
Gefahr
|
M-Clarity™ Qualitätsstufe = MQ100
107077-5G
SIAL107077-5GEA
266
EUR
SIAL107077-5G
1-Brom-2,3,4,5-tetrafluorbenzol, Sigma-Aldrich®
1-Bromo-2,3,4,5-tetrafluorobenzene
Formel:
C₆HBrF₄ Molecular Weight: 228,97 g/mol |
MDL:
MFCD00017900 CAS-Nummer: 1074-91-5 EINECS: 214-048-7 UN: 1993 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...