N-Succinimidyl 3-(Acetylthio)propionate (SATP), Sigma-Aldrich®

Lieferant: Merck

Synonyme: SATP

10859-50MG
SIAL10859-50MGEA 577 EUR
SIAL10859-50MG
N-Succinimidyl 3-(Acetylthio)propionate (SATP), Sigma-Aldrich®
SATP (N-Succinimidyl 3-(Acetylthio)propionate)

3-(Acetylthio)propionic acid N-succinimidyl ester causes tip functionalization and surface modification of antibody on atomic force microscope tips .


3-(Acetylthio)propionic acid N-succinimidyl ester may be used in oligosaccharide synthesis. It was used in binding antibody and ligand to atomic force microscope tips.
Conditionnement


Reagent for the modification of peptides and proteins: thionylation

Formel: C₉H₁₁NO₅S
Molecular Weight: 245,25 g/mol
MDL: MFCD00133023
CAS-Nummer: 84271-78-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR