Glutardialdehyd(dinatriumhydrogensulfit), Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

108790-50G
SIAL108790-50GEA 30.7 EUR
SIAL108790-50G
Glutardialdehyd(dinatriumhydrogensulfit), Sigma-Aldrich®
Glutardialdehyd(dinatriumhydrogensulfit)
Formel: C₅H₁₀Na₂O₈S₂
Molecular Weight: 308,24 g/mol
CAS-Nummer: 7420-89-5
EINECS: 231-043-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR