2-Chlorphenylhydraziniumchlorid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-Chlorphenylhydrazin Hydrochlorid, 2-Chlorphenylhydrazinhydrochlorid, 1-(2-Chlorphenyl)hydrazin Hydrochlorid

M-Clarity™ Qualitätsstufe = MQ100

109509-25G
SIAL109509-25GEA 248 EUR
SIAL109509-25G
2-Chlorphenylhydraziniumchlorid, Sigma-Aldrich®
2-Chlorphenylhydraziniumchlorid
Formel: C₆H₈Cl₂N₂
Molecular Weight: 179,05 g/mol
MDL: MFCD00012928
CAS-Nummer: 41052-75-9
EINECS: 255-194-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR