Antimon(V)sulfid, Sigma-Aldrich®

Lieferant: Merck
Achtung

Synonyme: di-Antimonpentasulfid, Antimonpentasulfid

M-Clarity™ Qualitätsstufe = MQ100

11133-500G-R
SIAL11133-500G-REA 223 EUR
SIAL11133-500G-R
Antimon(V)sulfid, Sigma-Aldrich®
Antimon(V)sulfid
Formel: S₅Sb₂
Molecular Weight: 403,85 g/mol
Dichte: 4,12 g/cm³ (20 °C)
MDL: MFCD00016321
CAS-Nummer: 1315-04-4
EINECS: 215-255-5
UN: 3178
ADR: 4.1,II
Merck Index: 12,00738

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR