3-Ethyl-3-pentanol, Sigma-Aldrich®

Lieferant: Merck

111570-50G
SIAL111570-50GEA 167 EUR
SIAL111570-50G
3-Ethyl-3-pentanol, Sigma-Aldrich®
3-Ethyl-3-pentanol
Formel: C₇H₁₆O
Molecular Weight: 116,2 g/mol
MDL: MFCD00004484
CAS-Nummer: 597-49-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR