Ethylcinnamat ≥99%, Sigma-Aldrich®
Lieferant: Merck
|
Achtung
|
Synonyme:
3-Phenylacrylsäureethylester, Cinnamsäureethylester, Zimtsäureethylester, 3-Phenylpropensäureethylester
M-Clarity™ Qualitätsstufe = MQ100
112372-100G
SIAL112372-100GEA
25
EUR
SIAL112372-100G
Ethylcinnamat ≥99%, Sigma-Aldrich®
Ethylcinnamat
Formel:
C₁₁H₁₂O₂ Molecular Weight: 176,21 g/mol |
MDL:
MFCD00009189 CAS-Nummer: 103-36-6 EINECS: 203-104-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...