4'-Chlor-2,2,2-trifluoracetophenon, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1-(4-Chlorophenyl)-2,2,2-trifluoroethanone

M-Clarity™ Qualitätsstufe = MQ100

112860-10G
SIAL112860-10GEA 181 EUR
SIAL112860-10G
4'-Chlor-2,2,2-trifluoracetophenon, Sigma-Aldrich®
4'-Chlor-2,2,2-trifluoracetophenon
Formel: C₈H₄ClF₃O
Molecular Weight: 208,5700 g/mol
MDL: MFCD00018686
CAS-Nummer: 321-37-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR