(3,4,5-Trimethoxyphenyl)acetonitril, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3,4,5-Trimethoxyphenylacetonitrile

M-Clarity™ Qualitätsstufe = MQ100

113360-10G
SIAL113360-10GEA 98.3 EUR
SIAL113360-10G
(3,4,5-Trimethoxyphenyl)acetonitril, Sigma-Aldrich®
(3,4,5-Trimethoxyphenyl)acetonitril
Formel: C₁₁H₁₃NO₃
Molecular Weight: 207,23 g/mol
MDL: MFCD00001912
CAS-Nummer: 13338-63-1
EINECS: 236-388-5
UN: 3439
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR