Chlor(dimethyl)phenylsilan, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

113379-25G 113379-5G
SIAL113379-25GEA 186 EUR
SIAL113379-25G SIAL113379-5G
Chlor(dimethyl)phenylsilan, Sigma-Aldrich®
Chlor(dimethyl)phenylsilan
Formel: C₈H₁₁ClSi
MDL: MFCD00000499
CAS-Nummer: 768-33-2
EINECS: 212-193-0
UN: 2987
ADR: 8,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR