HATU (O-(7-Aza-1H-benzotriazol-1-yl)-N,N,N',N'-tetramethyluroniumhexafluorophosphat), Sigma-Aldrich®

Lieferant: Merck

Synonyme: HATU, 1-[Bis(dimethylamino)methylene]-1H-1,2,3-triazolo[4,5-b]pyridinium 3-oxide, hexafluorophosphate

11373-1G 11373-25G 11373-5G
SIAL11373-1GEA 30.6 EUR
SIAL11373-1G SIAL11373-25G SIAL11373-5G
HATU (O-(7-Aza-1H-benzotriazol-1-yl)-N,N,N',N'-tetramethyluroniumhexafluorophosphat), Sigma-Aldrich®
HATU (O-(7-Aza-1H-benzotriazol-1-yl)-N,N,N',N'-tetramethyluroniumhexafluorophosphat)
Formel: C₁₀H₁₅F₆N₆OP
MDL: MFCD00274639
CAS-Nummer: 148893-10-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR