HATU (O-(7-Aza-1H-benzotriazol-1-yl)-N,N,N',N'-tetramethyluroniumhexafluorophosphat), Sigma-Aldrich®
Lieferant: Merck
Synonyme:
HATU, 1-[Bis(dimethylamino)methylene]-1H-1,2,3-triazolo[4,5-b]pyridinium 3-oxide, hexafluorophosphate
11373-1G
11373-25G
11373-5G
SIAL11373-1GEA
30.6
EUR
SIAL11373-1G
SIAL11373-25G
SIAL11373-5G
HATU (O-(7-Aza-1H-benzotriazol-1-yl)-N,N,N',N'-tetramethyluroniumhexafluorophosphat), Sigma-Aldrich®
HATU (O-(7-Aza-1H-benzotriazol-1-yl)-N,N,N',N'-tetramethyluroniumhexafluorophosphat)
Formel:
C₁₀H₁₅F₆N₆OP |
MDL:
MFCD00274639 CAS-Nummer: 148893-10-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...