1-Aza-12-krone-4 (1,4,7-Trioxa-10-azacyclododecan), Sigma-Aldrich®

Lieferant: Merck

11378-1G
SIAL11378-1GEA 311 EUR
SIAL11378-1G
1-Aza-12-krone-4 (1,4,7-Trioxa-10-azacyclododecan), Sigma-Aldrich®
1-Aza-12-krone-4 (1,4,7-Trioxa-10-azacyclododecan)
Formel: C₈H₁₇NO₃
MDL: MFCD00134269
CAS-Nummer: 41775-76-2
EINECS: 255-549-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR