Ethylcyclobutancarboxylat, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

113867-25G
SIAL113867-25GEA 155 EUR
SIAL113867-25G
Ethylcyclobutancarboxylat, Sigma-Aldrich®
Ethylcyclobutancarboxylat
Formel: C₇H₁₂O₂
Molecular Weight: 128,1690 g/mol
MDL: MFCD00001321
CAS-Nummer: 14924-53-9
EINECS: 238-995-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR