(2-Chlorethoxy)benzol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-Phenoxyethylchlorid, β-Chlorphenetol, (2-Chlorethyl)phenylether, ω-Chlorphenetol

114278-10ML
SIAL114278-10MLEA 58.5 EUR
SIAL114278-10ML
(2-Chlorethoxy)benzol, Sigma-Aldrich®
(2-Chlorethoxy)benzol
Formel: C₆H₅OCH₂CH₂Cl
Molecular Weight: 156,61 g/mol
MDL: MFCD00018946
CAS-Nummer: 622-86-6
EINECS: 210-757-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR