1,1-Diphenylhydrazin Hydrochlorid, Sigma-Aldrich®

Lieferant: Merck

114596-25G
SIAL114596-25GEA 110 EUR
SIAL114596-25G
1,1-Diphenylhydrazin Hydrochlorid, Sigma-Aldrich®
1,1-Diphenylhydrazin Hydrochlorid
Formel: C₁₂H₁₃ClN₂
MDL: MFCD00012950
CAS-Nummer: 530-47-2
EINECS: 208-481-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR