1,1,3,3-Tetramethyl-2-thioharnstoff, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Tetramethylthioharnstoff, Tetramethyl-2-thioharnstoff, N,N,N',N'-Tetramethylthioharnstoff
M-Clarity™ Qualitätsstufe = MQ100
115169-25G
SIAL115169-25GEA
74.6
EUR
SIAL115169-25G
1,1,3,3-Tetramethyl-2-thioharnstoff, Sigma-Aldrich®
1,1,3,3-Tetramethyl-2-thioharnstoff
Formel:
C₅H₁₂N₂S Molecular Weight: 132.23 g/mol Siedepunkt: 245 Schmelzpunkt: 74…76 °C |
MDL:
MFCD00008324 CAS-Nummer: 2782-91-4 EINECS: 220-488-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...