4-Aminonicotinsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4-Aminopyridin-3-carbonsäure, 4-Amino-3-pyridincarbonsäure

11585-5G-F
SIAL11585-5G-FEA 69.6 EUR
SIAL11585-5G-F
4-Aminonicotinsäure, Sigma-Aldrich®
4-Aminonicotinsäure
Formel: C₆H₆N₂O₂
Molecular Weight: 138,13 g/mol
MDL: MFCD00234183
CAS-Nummer: 7418-65-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR