2,6-Dinitrobenzaldehyd, Sigma-Aldrich®

Lieferant: Merck
Achtung

117501-1G
SIAL117501-1GEA 234 EUR
SIAL117501-1G
2,6-Dinitrobenzaldehyd, Sigma-Aldrich®
2,6-Dinitrobenzaldehyde
Formel: C₇H₄N₂O₅
Molecular Weight: 196,12 g/mol
Lagertemperatur: Kühlschrank
CAS-Nummer: 606-31-5
EINECS: 210-113-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR