2,6-Dichlor-3-nitrotoluol, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
1,3-Dichloro-2-methyl-4-nitrobenzene
M-Clarity™ Qualitätsstufe = MQ200
117536-25G
SIAL117536-25GEA
83.5
EUR
SIAL117536-25G
2,6-Dichlor-3-nitrotoluol, Sigma-Aldrich®
2,6-Dichlor-3-nitrotoluol
Formel:
C₇H₅Cl₂NO₂ Molecular Weight: 206,03 g/mol Schmelzpunkt: 53 °C |
MDL:
MFCD00024185 CAS-Nummer: 29682-46-0 EINECS: 249-776-4 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...