Cyclopentadienylmolybdäntricarbonyl dimer, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ100
117609-1G
SIAL117609-1GEA
50.7
EUR
SIAL117609-1G
Cyclopentadienylmolybdäntricarbonyl dimer, Sigma-Aldrich®
Cyclopentadienylmolybdäntricarbonyl dimer
Formel:
C₁₆H₁₀Mo₂O₆ |
MDL:
MFCD00009947 CAS-Nummer: 12091-64-4 EINECS: 235-156-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...