2-[Ethyl(phenyl)amino]ethanol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: N-Ethyl-N-(2-hydroxyethyl)aniline, Ethylphenylethanolamine, 2-(Ethyl(phenyl)amino)ethanol, N-Ethyl-N-2-ethanolaniline

M-Clarity™ Qualitätsstufe = MQ100

117684-100G
SIAL117684-100GEA 43.8 EUR
SIAL117684-100G
2-[Ethyl(phenyl)amino]ethanol, Sigma-Aldrich®
2-(N-Ethylanilino)ethanol
Formel: C₁₀H₁₅NO
Molecular Weight: 165,24 g/mol
MDL: MFCD00020575
CAS-Nummer: 92-50-2
EINECS: 202-160-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR