2-Cyanphenylacetonitril, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-(Cyanomethyl)benzonitrile, α-Cyano-o-tolunitrile, Homophthalonitrile

M-Clarity™ Qualitätsstufe = MQ100

118133-25G
SIAL118133-25GEA 65.9 EUR
SIAL118133-25G
2-Cyanphenylacetonitril, Sigma-Aldrich®
2-Cyanphenylacetonitril
Formel: C₉H₆N₂
Molecular Weight: 142.16 g/mol
MDL: MFCD00001895
CAS-Nummer: 3759-28-2
EINECS: 223-167-3
UN: 3439
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR