3-(p-Tolyl)propionsäure, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

118265-10G
SIAL118265-10GEA 100 EUR
SIAL118265-10G
3-(p-Tolyl)propionsäure, Sigma-Aldrich®
3-(p-Tolyl)propionsäure
Formel: C₁₀H₁₂O₂
MDL: MFCD00016561
CAS-Nummer: 1505-50-6
EINECS: 216-132-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR