Methyl-4-fluorbenzoat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4-Fluorbenzoesäuremethylester

M-Clarity™ Qualitätsstufe = MQ100

120707-10G
SIAL120707-10GEA 128 EUR
SIAL120707-10G
Methyl-4-fluorbenzoat, Sigma-Aldrich®
Methyl-4-fluorbenzoat
Formel: C₈H₇FO₂
Molecular Weight: 154,14 g/mol
MDL: MFCD00017959
CAS-Nummer: 403-33-8
EINECS: 206-956-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR