Benz[d]isoxazol, Sigma-Aldrich®

Lieferant: Merck

12255-5ML-F
SIAL12255-5ML-FEA 124 EUR
SIAL12255-5ML-F
Benz[d]isoxazol, Sigma-Aldrich®
Benz[d]isoxazol
Formel: C₇H₅NO
Molecular Weight: 119,12 g/mol
MDL: MFCD00005852
CAS-Nummer: 271-95-4
EINECS: 205-983-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR