2-Amino-4-chlor-6-methylpyrimidin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4-Chloro-6-methylpyrimidin-2-amine

M-Clarity™ Qualitätsstufe = MQ100

122882-25G 122882-5G
SIAL122882-25GEA 88.3 EUR
SIAL122882-25G SIAL122882-5G
2-Amino-4-chlor-6-methylpyrimidin, Sigma-Aldrich®
2-Amino-4-chloro-6-methylpyrimidine
Formel: C₅H₆ClN₃
Molecular Weight: 143.58 g/mol
MDL: MFCD00006091
CAS-Nummer: 5600-21-5
EINECS: 227-018-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR