1-Benzyl-2-pyrrolidinon, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1-Benzylpyrrolidin-2-one

M-Clarity™ Qualitätsstufe = MQ100

123757-25G
SIAL123757-25GEA 59.9 EUR
SIAL123757-25G
1-Benzyl-2-pyrrolidinon, Sigma-Aldrich®
1-Benzyl-2-pyrrolidinon
Formel: C₁₁H₁₃NO
Molecular Weight: 175.23 g/mol
MDL: MFCD00003195
CAS-Nummer: 5291-77-0
EINECS: 226-131-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR