2-Chlor-4,6-dinitrophenol, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

124613-25G
SIAL124613-25GEA 216 EUR
SIAL124613-25G
2-Chlor-4,6-dinitrophenol, Sigma-Aldrich®
2-Chlor-4,6-dinitrophenol
Formel: C₆H₃ClN₂O₅
Molecular Weight: 218,55 g/mol
CAS-Nummer: 946-31-6
EINECS: 213-419-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR