2-Chlor-6-methylbenzonitril, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

125083-5G
SIAL125083-5GEA 105 EUR
SIAL125083-5G
2-Chlor-6-methylbenzonitril, Sigma-Aldrich®
2-Chlor-6-methylbenzonitril
Formel: C₈H₆ClN
Molecular Weight: 151,60 g/mol
MDL: MFCD00001782
CAS-Nummer: 6575-09-3
EINECS: 229-499-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR