2-Cyan-6-methylpyridin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 6-Methyl-2-pyridincarbonitril, 6-Cyan-2-picolin

M-Clarity™ Qualitätsstufe = MQ100

125385-5G
SIAL125385-5GEA 94.7 EUR
SIAL125385-5G
2-Cyan-6-methylpyridin, Sigma-Aldrich®
2-Cyano-6-methylpyridine
Formel: C₇H₆N₂
Molecular Weight: 118,14 g/mol
MDL: MFCD00190586
CAS-Nummer: 1620-75-3
EINECS: 216-588-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR