5-Chlor-2-mercaptobenzothiazol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 5-Chlorobenzo[d]thiazole-2(3H)-thione

M-Clarity™ Qualitätsstufe = MQ200

125571-5G
SIAL125571-5GEA 56.4 EUR
SIAL125571-5G
5-Chlor-2-mercaptobenzothiazol, Sigma-Aldrich®
5-Chloro-2-mercaptobenzothiazole
Formel: C₇H₄ClNS₂
Molecular Weight: 201,7 g/mol
MDL: MFCD00005783
CAS-Nummer: 5331-91-9
EINECS: 226-235-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR