1,2-Diphenylhydrazin, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ200

126721-25G
SIAL126721-25GEA 101 EUR
SIAL126721-25G
1,2-Diphenylhydrazin, Sigma-Aldrich®
1,2-Diphenylhydrazine
Formel: C₁₂H₁₂N₂
MDL: MFCD00003012
CAS-Nummer: 122-66-7
EINECS: 204-563-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR