N,N,N'-Triethylethylendiamin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Diethyl(2-ethylaminoethyl)amin, 1-(Diethylamino)-2-(ethylamino)ethan

127116-5G
SIAL127116-5GEA 51.2 EUR
SIAL127116-5G
N,N,N'-Triethylethylendiamin, Sigma-Aldrich®
N,N,N'-Triethylethylendiamin
Formel: C₈H₂₀N₂
Molecular Weight: 144,26 g/mol
Siedepunkt: 54 °C (1013 hPa)
Dichte: 0,804 g/cm³ (20 °C)
MDL: MFCD00009054
CAS-Nummer: 105-04-4
EINECS: 203-264-7
UN: 2734
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR