N,N,N'-Trimethylethylendiamin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1-(Dimethylamino)-2-(methylamino)ethan, Dimethyl(2-(methylamino)ethyl)amin

127124-25G 127124-5G
SIAL127124-25GEA 283 EUR
SIAL127124-25G SIAL127124-5G
N,N,N'-Trimethylethylendiamin, Sigma-Aldrich®
N,N,N'-Trimethylethylendiamin
Formel: C₅H₁₄N₂
Molecular Weight: 102.18 g/mol
Siedepunkt: 116
Dichte: 0.783 g/cm³ (20 °C)
MDL: MFCD00014874
CAS-Nummer: 142-25-6
EINECS: 205-529-2
UN: 2733
ADR: 3 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR