Diphenyliodoniumnitrat, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

127396-25G
SIAL127396-25GEA 122 EUR
SIAL127396-25G
Diphenyliodoniumnitrat, Sigma-Aldrich®
Diphenyliodonium nitrate
Formel: C₁₂H₁₀INO₃
Molecular Weight: 343,12 g/mol
MDL: MFCD00031704
CAS-Nummer: 722-56-5
EINECS: 211-962-8
UN: 3087
ADR: 5.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR