2-Amino-5-mercapto-1,3,4-thiadiazol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 5-Amino-1,3,4-thiadiazole-2-thiol

M-Clarity™ Qualitätsstufe = MQ200

127906-50G
SIAL127906-50GEA 59.2 EUR
SIAL127906-50G
2-Amino-5-mercapto-1,3,4-thiadiazol, Sigma-Aldrich®
2-Amino-5-mercapto-1,3,4-thiadiazole
Formel: C₂H₃N₃S₂
Molecular Weight: 133,2 g/mol
MDL: MFCD00003108
CAS-Nummer: 2349-67-9
EINECS: 219-078-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR