HBTU (O-(1H-Benzotriazol-1-yl)-N,N,N',N'-tetramethyluroniumhexafluorophosphat), Sigma-Aldrich®

Lieferant: Merck

Synonyme: HBTU, 1-(Bis(dimethylamino)methylene)-1H-benzo[d][1,2,3]triazole-1-ium 3-oxide hexafluorophosphate(V)

12804-1G-F 12804-25G-F 12804-5G-F
SIAL12804-1G-FEA 45.6 EUR
SIAL12804-1G-F SIAL12804-25G-F SIAL12804-5G-F
HBTU (O-(1H-Benzotriazol-1-yl)-N,N,N',N'-tetramethyluroniumhexafluorophosphat), Sigma-Aldrich®
HBTU (O-(1H-Benzotriazol-1-yl)-N,N,N',N'-tetramethyluroniumhexafluorophosphat)
Formel: C₁₁H₁₆F₆N₅OP
Molecular Weight: 379.2 g/mol
MDL: MFCD00075445
CAS-Nummer: 94790-37-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR