(Benzotriazol-1-yloxy)tri(1-pyrrolidinyl)phosphoniumhexafluorophosphat (PyBOP), Sigma-Aldrich®

Lieferant: Merck

Synonyme: PyBOP, ((1H-Benzo[d][1,2,3]triazol-1-yl)oxy)tri(pyrrolidin-1-yl)phosphonium hexafluorophosphate(V), 1H-Benzotriazol-1-yloxytri(1-pyrrolidinyl)phosphonium hexafluorophosphate

M-Clarity™ Qualitätsstufe = MQ200

12805-1G-F 12805-25G-F 12805-5G-F
SIAL12805-1G-FEA 71.5 EUR
SIAL12805-1G-F SIAL12805-25G-F SIAL12805-5G-F
(Benzotriazol-1-yloxy)tri(1-pyrrolidinyl)phosphoniumhexafluorophosphat (PyBOP), Sigma-Aldrich®
(Benzotriazol-1-yloxy)tri(1-pyrrolidinyl)phosphoniumhexafluorophosphat (PyBOP)
Formel: C₁₈H₂₈F₆N₆OP₂
MDL: MFCD00077411
CAS-Nummer: 128625-52-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR